VWR® PCR Plates, 96-Well
供应商: VWR Collection
82006-636
VWRI211-0262EA 5257 CNY
VWRI211-0262 VWRI211-0263 VWRI211-0264 VWRI211-0265 VWRI211-0266 VWRI211-0267 VWRI211-0268 VWRI211-0269 VWRI211-0298 VWRI211-0313 VWRI211-0283BC VWRU82006-636 VWRI732-3487 VWRI732-3488 VWRI732-3494
VWR® PCR Plates, 96-Well
微孔板 PCR板
VWR® 96-Well PCR Plates are compatible with most leading PCR and real-time PCR instruments.
  • Ultra-thin walls to optimize thermal transfer resulting in increased reaction efficiencies
  • Ultra-clear wells for easy visualization of small sample volumes
  • Made of virgin polypropylene which is naturally low-binding without the need to incorporate additives
  • Black alphanumerics are easy-to-read and simplifies sample identification.
  • All plates are certified free of RNAse, DNAse, and human DNA
Compatible with VWR® 8-strip flat caps (211-0350) and bubble caps (732-0More Product Information