多层粘尘垫, PureStep
供应商: VWR Collection
82031-676 87004-016 87004-040
VWRI115-0047EA 1237.33 CNY
VWRI115-0047 VWRI115-0048 VWRI115-0049 VWRI115-0050 VWRI115-0051 VWRI115-0052 VWRI115-0053 VWRI115-0054 VWRI115-0055 VWRI115-0056 VWRI115-0057 VWRI115-0058 VWRI115-0059 VWRI115-0060 VWRI115-0061 VWRI115-0062 VWRI115-0063 VWRI115-0064 VWRI115-0065 VWRI115-0066 VWRI115-0067 VWRI115-0068 VWRI115-0069 VWRI115-0070 VWRI115-0071 VWRI115-0072 VWRI115-0075 VWRI115-0076 VWRI115-0077 VWRI115-0078 VWRI115-0079 VWRI115-0080 VWRI115-0081 VWRI115-0082 VWRI115-0083 VWRI115-0084 VWRI115-0085 VWRI115-0086 VWRI115-0087 VWRI115-0088 VWRI115-0089 VWRI115-0090 VWRI115-0091 VWRI115-0092 VWRI115-0093 VWRI115-0073 VWRI115-0074 VWRI115-0167 VWRI115-0168 VWRI115-0169 VWRI115-1748 VWRU82031-676 VWRU87004-016 VWRI115-1774 VWRI115-1775 VWRI115-1776 VWRU87004-040
多层粘尘垫, PureStep
粘尘垫 粘合地板垫
30或60层粘尘垫可供选择,采用低密度聚乙烯制造。可清除鞋底和运输容器轮子上的碎屑,从而控制污染。一层变脏时,只要按照角落里的编号将其取下,露出新的下一层即可。
  • 抗菌剂可防止细菌和真菌生长
  • 防滑,低而稳固,单独的编号层
  • 可降低维护费和清洁费
  • Contains GMO-free, renewable plant-based material that reduces CO₂ emissions
  • Adhesive mats are also recyclable after use
颜色:透明、白色、灰色或蓝色.
Used mat layers, including top and bottom release liners, can be recycled after use using the Recycling solution for plastic packaging, Zero Waste Box™ from TerraCycle.