VWR® Basic, Bouffant Caps, Latex-Free
供应商: VWR Collection
89107-768 89131-620 414005-000 414005-001 89131-622
VWRI113-1459EA 594.31 CNY
VWRI113-1459 VWRI113-1460 VWRU89107-768 VWRU89131-620 VWRU414005-000 VWRU414005-001 VWRU89131-622 VWRI113-8762 VWRI113-8877 VWRI113-8887 VWRI113-8886 VWRI113-8888 VWRI113-8874 VWRI113-8875 VWRI113-8876 VWRI113-8879 VWRI113-8882 VWRI113-8880 VWRI113-8881 VWRI113-8883 VWRI113-8878 VWRI113-8884 VWRI113-8885
VWR® Basic, Bouffant Caps, Latex-Free
Disposable Garments
These disposable caps are made from a non woven, spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates, in non hazardous environments.
  • Free from natural rubber latex
  • Available in a wide range of colours
Weight: 10 g/m²
Packaging: 100 pieces per bag, 10 bags per case.
现在订购
规格
Environmentally Preferable
Gender
Garment application