洁净室鞋套,防滑,VWR最大防护型,SF
供应商: VWR Collection
414004-857
VWRI113-1374EA 2411 CNY
VWRI113-1374 VWRI113-1375 VWRI113-1373 VWRI113-1372 VWRI113-1371 VWRI113-1370 VWRI113-1369 VWRI113-1376 VWRU414004-857 VWRI114-9140
洁净室鞋套,防滑,VWR最大防护型,SF
Disposable Garments
VWR®最大防护型SF鞋套在整个VWR®服装系列中最洁净、最耐用。采用纺粘丙纶织物(SPP),带透气性聚乙烯薄膜,这些材料具有最佳的屏障保护、洁净度和舒适性。
  • Non skid and anti-static
  • Impervious to fluids
  • Superior friction coefficient to ensure maximum traction
  • Maximum skid resistance on wet and smooth surfaces
  • Clean, low linting fabric
  • 60 g laminated fabric with PVC sole
  • Particle shedding (IEST-RP-CC003.3) level 1 E
  • Sterile version (article; 114-9140)
  • Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
在经ISO认证的设施中,在严格的过程控制下进行测试和制造。通过独立的实验室测试进行批次控制和验证。
长度:359 mm (U*), 292 mm (M), 324 mm (L), 381 mm (XL)
Caution: *UMore Product Information