VWR®, Clamps, Three Pronged, with Flexible Rod
供应商: VWR Collection
VWRF08303.157EA 0 CNY
VWRF08303.157
VWR®, Clamps, Three Pronged, with Flexible Rod
夹具 通用夹具
Laboral aluminium alloy.
  • Jaws covered in vinyl to protect clamped items
  • Clamp rod diameter: 8 mm; rod length: 200 mm