VWR®, Clamps
供应商: VWR Collection
VWRF08235.915EA 0 CNY
VWRF08235.915 VWRF08257.556
VWR®, Clamps
夹具 通用夹具
Laboral aluminium alloy.
  • Round jaws covered in vinyl to protect clamped items
  • One easy action knurled screw for securing