溴化乙啶 solution_stock, Biotechnology Grade

供应商: VWR Chemicals

Synonyms: EtBr, 胡米溴铵, 3,8-二氨基-5-乙基-6-苯基菲啶溴化物

VWRCX328-10MLEA 765 CNY
VWRCX328-10ML
溴化乙啶 solution_stock, Biotechnology Grade
溴化乙啶

A 10mg/ml solution of Ethidium Bromide in specially prepared deionized water. Ethidium bromide solution is a fluorescent stain for visualising nucleic acids in solution or in electrophoresis gels.


At a concentration of 0,5 μg/ml, ethidium bromide is useful in agarose or acrylamide gels for band detection. It is used to detect as little as 10 ng of nucleic acid. At higher concentrations (0,5 - 1,0 μg/ml), it may be used to facilitate purification of DNA in a caesium chloride gradient.

分子式: C₂₁H₂₀BrN₃
分子量: 394.31 g/mol
沸点: 72
存储条件: Ambient
MDL编号: MFCD00011724
CAS号: 1239-45-8
Merck Index: 13,04751

现在订购

产品技术规格

DNase none detected
Moisture <= 8%
Purity (Dry Basis) > = 98%
RNase none detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR