VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline), USP
供应商: VWR Chemicals
VWRCK813-500MLEA 1117 CNY
VWRCK813-500ML
VWR® UltraPure, Buffer, PBS solution 10X concentrate (phosphate buffered saline), USP
缓冲(液) 组织培养缓冲液
  • No DNases, RNases or proteases detected
  •  
Product tested according to USP specification
pH @ 25 °C (1:10 water)    6,6 to 7,2