乙酸铵 5mM water, HiPerSolv CHROMANORM®, PFAS grade eluent, LC-MS

供应商: VWR Chemicals

Synonyms: Acetic acid ammonium salt

VWRC92500.290EA 0 CNY
VWRC92500.290
乙酸铵 5mM water, HiPerSolv CHROMANORM®, PFAS grade eluent, LC-MS
乙酸铵

VWR® PFAS grade LC-MS solvents undergo a rigorous use test to ensure suitability for analysis of poly- and perfluoroalkyl substances resulting in an ultra low background of PFAS analytes. Using complementary methods a broad range of PFAS analytes are quantified to their detection limits by LC-MS.


  • Specifically tested for PFAS analysis
  • Ultra low background solvent for LC-MS analysis of PFAS (poly- and perfluoroalkyl substances)

分子式: H₃CCOONH₄
分子量: 77.08 g/mol
溶点: 114
闪点: 136
存储条件: Ambient
MDL编号: MFCD00013066
CAS号: 631-61-8
EINECS: 231-791-2
Merck Index: 13,00495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd160345483553318b94053de396335a9b18425f&vid=c6375b539008d6b43f50408d30b61261444faebc&prid=web&clid=SAPDAM

现在订购

产品技术规格

Nominal concentration (V/V) 4.75 - 5.25 mM
Ca (Calcium) max. 0.2 ppm
K (Potassium) max. 0.2 ppm
Mg (Magnesium) max. 0.2 ppm
Na (Sodium) max. 0.5 ppm
Suitability for PFAS analysis passes test
manufactured and filled under nitrogen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR