VWR® Fixing Solution
供应商: VWR Chemicals
VWRC87970.260EA 2072 CNY
VWRC87970.260
VWR® Fixing Solution
染料 显微和染色试剂
Fixing solution with 0,02% formaldehyde in methanol 50% (v/v)).
订购须知 Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.