氯化钠 20% in aqueous solution Reag. Ph. Eur. 1079502

供应商: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC87931.290EA 2109 CNY
VWRC87931.290
氯化钠 20% in aqueous solution Reag. Ph. Eur. 1079502
氯化钠

订购须知 Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

分子式: NaCl
分子量: 58.44 g/mol
沸点: 1413
溶点: 801
密度: 1…1.2 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003477
CAS号: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

现在订购

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR