1,4-二氧六环 0,05% aqueous solution Reag. Ph. Eur. 1032002

供应商: VWR Chemicals

Synonyms: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide

VWRC87821.180EA 3796 CNY
VWRC87821.180
1,4-二氧六环 0,05% aqueous solution Reag. Ph. Eur. 1032002
1,4-二氧六环

订购须知 Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

分子式: C₄H₈O₂
分子量: 88.11 g/mol
沸点: 101.1
溶点: 11.8
闪点: 12
MDL编号: MFCD00006571
CAS号: 123-91-1
EINECS: 204-661-8
UN号: 1165
ADR: 3 II
Merck Index: 13,03330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR