VWR® AVS TITRINORM, Titanium Standard Solution, 1000 mg/l Ti in 5% Nitric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
供应商: VWR Chemicals
VWRC86717.260EA 974 CNY
VWRC86717.260 VWRC86717.180
VWR® AVS TITRINORM, Titanium Standard Solution, 1000 mg/l Ti in 5% Nitric Acid with Hydrofluoric Acid (max. 1%), Standard for AAS
标准品 Cationic Standards Titanium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%