VWR® AVS TITRINORM, Scandium Standard Solution, 1000 mg/L Sc in dil. Nitric Acid, Standard for AAS
供应商: VWR Chemicals
VWRC86708.180EA 2691 CNY
VWRC86708.180 VWRC86708.260
VWR® AVS TITRINORM, Scandium Standard Solution, 1000 mg/L Sc in dil. Nitric Acid, Standard for AAS
标准品 Cationic Standards Scandium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%