VWR® AVS TITRINORM, Germanium Standard Solution, 1000 mg/L Ge in 5% Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
供应商: VWR Chemicals
VWRC86680.180EA 1011 CNY
VWRC86680.180 VWRC86680.260
VWR® AVS TITRINORM, Germanium Standard Solution, 1000 mg/L Ge in 5% Nitric Acid with hydrofluoric Acid (max. 1%) AVS TITRINORM Standard for AAS
标准品 Cationic Standards Germanium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%