VWR® AVS TITRINORM, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid AVS TITRINORM Standard for AAS
供应商: VWR Chemicals
VWRC86676.180EA 1469 CNY
VWRC86676.180 VWRC86676.260
VWR® AVS TITRINORM, Europium Standard Solution, 1000 mg/L Eu in dilute Nitric Acid AVS TITRINORM Standard for AAS
标准品 Cationic Standards Europium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%