VWR® AVS TITRINORM, Boron Standard Solution, 1000 mg/L B in Water, Standard for AAS
供应商: VWR Chemicals
VWRC86663.260EA 928 CNY
VWRC86663.260 VWRC86663.180
VWR® AVS TITRINORM, Boron Standard Solution, 1000 mg/L B in Water, Standard for AAS
标准品 Cationic Standards Boron Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%