VWR®, Density standards referenced at 15 °C
供应商: VWR Chemicals
VWRC85140.180EA
2064
CNY
VWRC85140.180
VWRC85141.180
VWRC85142.180
VWRC85143.180
VWRC85144.180
VWRC85148.180
VWRC85149.180
VWRC85150.180
VWRC85151.180
VWRC85152.180
VWRC85146.180
VWRC85147.180
VWRC85153.180
VWRC85155.180
VWRC85157.180
VWRC85158.180
VWRC85159.180
VWRC85160.180
VWRC85163.180
VWRC85165.180
VWR®, Density standards referenced at 15 °C
标准品
Density Standards
These products can be used as calibration standards for density measurements by pycnometric, vibrational or hydrometer-based techniques.
- High accuracy across an extensive density range (0,6960 to 3,1140 g/ml) at 15 °C
- No toxic heavy metals used in any formulation
- Can be used with any brand or type of density measuring instrument
- Supplied in a high quality, tamper-proof, amber glass bottle
These standards are prepared gravimetrically (solute and solvent) using OIML traceable weights. The density values of the standards are established using fully calibrated reference pycnometers.
Certifications: In accordance with ASTM D1480-12 for testing of density or relative density. A Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.
规格
Environmentally Preferable |