30% active coumpound, Snapsil RE20 Foam_control

供应商: VWR Chemicals

Synonyms: Anti-foaming agent

VWRC84538.290EA 750 CNY
VWRC84538.290
30% active coumpound, Snapsil RE20 Foam_control
Silicone antifoam

现在订购

产品技术规格

Active matter content 29.0 - 33.0 %
Density (25/4) 0.950 - 1.100
pH (20°C) 5.0 - 9.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR