VWR®, Turbidity standards Set
供应商: VWR Chemicals
VWRC84209.600EA 7586 CNY
VWRC84209.600
VWR®, Turbidity standards Set
标准品 浊度标准
Turbidity standards in kit form or alternatively as formazine stock solutions which allows users to create their own defined standards.
  • Set of standards designed for use with turbidity meters TIR 200 and TIR 210 with a measuring range from 0,01 to 1100 NTU
  • Ideal for units that measure scattered light at an angle of 90 0, as stipulated in EN ISO 7027
Certifications: ISO 7027
Delivery information: Kit supplied in handy case