VWR® ARISTAR®, ICP Multi-Element Calibration Standard 3 in dilute Nitric Acid
供应商: VWR Chemicals
VWRC456472JEA 13986 CNY
VWRC456472J
VWR® ARISTAR®, ICP Multi-Element Calibration Standard 3 in dilute Nitric Acid
标准品 Cationic Standards Multi-Element Standards
In 2% nitric acid solution.
4 Elements: 1000 mg/L Ba, Ca, Mg, Sr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.