VWR® ARISTAR®, Praseodymium Standard Solution, 1000 mg/l Pr in dil. Nitric Acid (from Pr6O11), Standard for ICP
供应商: VWR Chemicals
VWRC455842LEA 2024 CNY
VWRC455842L
VWR® ARISTAR®, Praseodymium Standard Solution, 1000 mg/l Pr in dil. Nitric Acid (from Pr6O11), Standard for ICP
标准品 Cationic Standards Praseodymium Standards
Pr₆O₁₁ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.