VWR® ARISTAR®, Nickel Standard Solution, 1000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
供应商: VWR Chemicals
VWRC455722EEA 1638 CNY
VWRC455722E VWRC455724G
VWR® ARISTAR®, Nickel Standard Solution, 1000 mg/L Ni in dil. Nitric Acid (from Ni), Standard for ICP
标准品 Cationic Standards Nickel Standards
Ni in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.