VWR® ARISTAR®, Indium Standard Solution, 1000 mg/L In in dilute Nitric Acid (from In), Standard for ICP
供应商: VWR Chemicals
VWRC455484JEA 3952 CNY
VWRC455484J VWRC455482H
VWR® ARISTAR®, Indium Standard Solution, 1000 mg/L In in dilute Nitric Acid (from In), Standard for ICP
标准品 Cationic Standards Indium Standards
In in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.