VWR®, Hafnium standard solution, 10,000 mg/L Hf in dil. nitric acid with hydrofluoric acid (max. 1%) (from HfO2) ARISTAR® standard for ICP
供应商: VWR Chemicals
VWRC455452BEA 6506 CNY
VWRC455452B VWRC455454D
VWR®, Hafnium standard solution, 10,000 mg/L Hf in dil. nitric acid with hydrofluoric acid (max. 1%) (from HfO2) ARISTAR® standard for ICP
标准品 Cationic Standards Hafnium Standards
HfO₂ in HNO₃ (2 to 5%) with traces HF.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.