VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
供应商: VWR Chemicals
VWRC455262UEA 1638 CNY
VWRC455262U VWRC455264W
VWR® ARISTAR®, Cobalt Standard Solution, for ICP, 1000 mg/L Co in dil. Nitric Acid (from Co)
标准品 Cationic Standards Cobalt Standards
Co in HNO₃ 2 to 5%.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.