VWR® ARISTAR®, Bismuth Standard Solution, 10000 mg/L Bi in dil. Nitric Acid (from Bi), Standard for ICP
供应商: VWR Chemicals
VWRC455112HEA 3133 CNY
VWRC455112H VWRC455114J
VWR® ARISTAR®, Bismuth Standard Solution, 10000 mg/L Bi in dil. Nitric Acid (from Bi), Standard for ICP
标准品 Cationic Standards Bismuth Standards
Bi in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.