VWR® ARISTAR®, Antimony Standard Solutions for ICP
供应商: VWR Chemicals
VWRC455022GEA 1638 CNY
VWRC455022G VWRC455024Y
VWR® ARISTAR®, Antimony Standard Solutions for ICP
标准品 Cationic Standards Antimony Standards
Sb in HNO₃ 2% tr. tartaric acid.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery information: Supplied with certificate of analysis.