2,2,4-三甲基戊烷 ≥98%, TECHNICAL

供应商: VWR Chemicals
Danger

Synonyms: 异辛烷

VWRC28775.290EA 473 CNY
VWRC28775.290 VWRC28775.368 VWRC28775.461
2,2,4-三甲基戊烷 ≥98%, TECHNICAL
2,2,4-三甲基戊烷
分子式: (CH₃)₂CHCH₂C(CH₃)₃
分子量: 114.23 g/mol
沸点: 99.2 °C (1013 hPa)
溶点: –107 °C
密度: 0.692 g/cm³ (20 °C)
闪点: –12 °C
存储条件: Ambient
MDL编号: MFCD00008943
CAS号: 540-84-1
EINECS: 208-759-1
UN号: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM

现在订购

产品技术规格

Assay Min. 98 %
Refractive index (20°C) 1.391 - 1.393

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR