高氯酸钠一水合物 98.0-102.0%, AnalaR NORMAPUR® analytical reagent

供应商: VWR Chemicals
Danger

Synonyms: Perchloric acid sodium salt

VWRC27988.232EA 1571 CNY
VWRC27988.232
高氯酸钠一水合物 98.0-102.0%, AnalaR NORMAPUR® analytical reagent
高氯酸钠一水合物
分子式: NaClO₄·H₂O
分子量: 140.46 g/mol
溶点: 130
密度: 2.02 g/cm³ (25 °C)
存储条件: Ambient
MDL编号: MFCD00149164
CAS号: 7791-07-3
EINECS: 231-511-9
UN号: 1502
ADR: 5.1 II
Merck Index: 12,08798

现在订购

产品技术规格

Assay 98,0% – 102,0%
pH (20 °C; 5%) 4.5 - 7.0
Heavy Metals (as Pb) Max. 5 ppm
Total N (Nitrogen) Max. 5 ppm
Cl (Chloride) Max. 30 ppm
SO4 (Sulphate) Max. 20 ppm
Ca (Calcium) Max. 20 ppm
Fe (Iron) Max. 3 ppm
K (Potassium) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR