戊烷 99, GPR RECTAPUR®

供应商: VWR Chemicals
Danger

Synonyms: Pentane

VWRC26206.291EA 455 CNY
VWRC26206.291 VWRC26206.325 VWRC26206.360 VWRC26206.460
戊烷 99, GPR RECTAPUR®
戊烷
分子式: H₃C(CH₂)₃CH₃
分子量: 72.15 g/mol
沸点: 35…36 °C (1013 hPa)
溶点: –130 °C
密度: 0.63 g/cm³ (20 °C)
闪点: –40 °C
存储条件: Ambient
MDL编号: MFCD00009498
CAS号: 109-66-0
EINECS: 203-692-4
UN号: 1265
ADR: 3 II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

现在订购

产品技术规格

Assay Min. 99 %
Boiling point 35 - 37 °C
Density (20/4) 0.625 - 0.632
Evaporation residue Max. 50 ppm

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR