硝酸镍(II) 96, GPR RECTAPUR®

供应商: VWR Chemicals
Danger

VWRC25871.292EA 1966 CNY
VWRC25871.292
硝酸镍(II) 96, GPR RECTAPUR®
硝酸镍(II)
分子式: Ni(NO₃)₂·6H₂O
分子量: 290.79 g/mol
沸点: 137
溶点: 56.7
密度: 2.05 g/cm³ (20 °C)
MDL编号: MFCD00149805
CAS号: 13478-00-7
EINECS: 236-068-5
UN号: 2725
ADR: 5.1 III
Merck Index: 12,06596

现在订购

产品技术规格

Assay Min. 96 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.05 %
Cd (Cadmium) Max. 25 ppm
Cu (Copper) Max. 25 ppm
Pb (Lead) Max. 25 ppm
Zn (Zinc) Max. 25 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR