Murexide analytical reagent

供应商: VWR Chemicals

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

VWRC25717.120EA 326 CNY
VWRC25717.120
Murexide analytical reagent
Murexide
分子式: C₈H₈N₆O₆
分子量: 284.19 g/mol
溶点: 300
存储条件: Ambient
MDL编号: MFCD00012777
CAS号: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330

现在订购

产品技术规格

Suited for complexometry indicator Passes test

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR