硫酸铵铝 ≥98%, GPR RECTAPUR®

供应商: VWR Chemicals
Warning

Synonyms: 硫酸铝铵, 铵明矾

VWRC21083.292EA 529 CNY
VWRC21083.292
硫酸铵铝 ≥98%, GPR RECTAPUR®
硫酸铵铝
分子式: AlNH₄(SO₄)₂·12H₂O
分子量: 453.33 g/mol
沸点: 200
溶点: 93
密度: 1.64 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00051042
CAS号: 7784-26-1
EINECS: 232-055-3
Merck Index: 13,00325

现在订购

产品技术规格

Assay Min. 98 %
Heavy metals (as Pb) Max. 50 ppm
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 10 ppm

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR