VWR® Pipette Tips

供应商: VWR Collection
VWRA612-6726EA 1930.11 CNY
VWRA612-6726 VWRA612-6727 VWRA612-6728 VWRA612-6729 VWRA612-6730 VWRA612-6731 VWRA612-6732 VWRA612-6733 VWRA612-6734 VWRA612-6735 VWRA612-6736 VWRA612-6737 VWRA612-6738 VWRA612-6739 VWRA612-6740 VWRA612-6741 VWRA612-6742 VWRA612-6743 VWRA612-6744 VWRA612-6745 VWRA612-6746 VWRA612-6747 VWRA612-6748 VWRA612-6749 VWRA612-6750 VWRA612-6751 VWRA612-6752 VWRA612-6753 VWRA612-6754 VWRA612-6755 VWRA612-6756 VWRA612-6757 VWRA612-6758 VWRA612-6759 VWRA612-6760 VWRA612-6761 VWRA612-6762 VWRA612-6763 VWRA612-6764 VWRA612-6765 VWRA612-6766 VWRA612-6767 VWRA612-6768 VWRA612-6792 VWRA612-6799 VWRA612-6800
VWR® Pipette Tips
吸头
These VWR® pipette tips are designed for precise liquid transfer.

  • Fully autoclavable to 121 °C
  • Colour-coded racks for easy volume identification
  • Tips with PP filter are also available
  • DNase-/RNase-free and non-pyrogenic
  • Every rack or case is printed with a batch number for traceability
  • Sterile tips are sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf Life: 3 years after month of production

Good quality consumable products for micropipettors. Preferred accessories for most brand micropipettors.

Certifications: Manufactured in a class 100 000More Product Information
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR