Powder funnel with short, broad stem
供应商: VITLAB
71194
VITL71194EA 0 CNY
VITL71194
Powder funnel with short, broad stem
漏斗 粉末漏斗
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.