Measuring cylinder, tall form, class B, PP
供应商: VITLAB
646941 647941 648941 649941 651941 652941 653941
VITL646941EA 0 CNY
VITL646941 VITL647941 VITL648941 VITL649941 VITL651941 VITL652941 VITL653941
Measuring cylinder, tall form, class B, PP
量筒 Measuring Cylinders
PP, translucent, tall form, class B.
  • Calibrated to 'In'
  • Hexagonal base, ring mark on the principal points
  • Thermal stresses of up to +80 °C do not result in the tolerance limit being permanently exceeded
Certifications: DIN 12681 B / ISO 6706