Tubing, Tygon® A-60-G
供应商: Saint Gobain Life Sciences
AFL00029-S
VERNAFL00029-SEA 0 CNY
VERNAFL00029-S
Tubing, Tygon® A-60-G
胶管 Transfer Tubing
Thermoplastic elastomer, black.
  • Long lasting, weatherproof, good abrasion resistance and exceptional resistance to fatigue caused by continual flexing
  • Excellent chemical resistance
  • Low gas permeability compared with rubber tubing
Versatile tubing for industrial and laboratory applications. Heat- and ozone-resistant. Excellent resistance to inorganic liquids (acids and bases). Ideal for use in peristaltic pumps.
Temperature resistance: −60 to +135 °C
Hardness (Shore durometer A): 61
Density: 0,98