Safety helmets, uvex pheos B-WR
供应商: UVEX
9772.030
UVEX9772.030EA 0 CNY
UVEX9772.030
Safety helmets, uvex pheos B-WR
头盔 Safety Helmets
These ventilated helmets provide protection even in extremely low temperatures (down to −30 °C). They also provide protection against splashes of molten metal.
  • Three variable air vents aid airflow and help regulate the temperature within the helmet, making it more comfortable to wear
  • Adapter systems for attaching uvex visors, uvex earmuffs and lamps
  • Suspension harness with rotating ratchet wheel allows the width size to be adjusted for an individual fit
Size: 520 to 610 mm
EN 397, low temperature (–30 °C), and molten metal
Certifications: EN 397,More Product Information