Chemical resistant gloves, uvex profabutyl B-05R
供应商: UVEX
6094911 6094910 6094907 6094908 6094909
UVEX6094911EA 0 CNY
UVEX6094911 UVEX6094910 UVEX6094907 UVEX6094908 UVEX6094909
Chemical resistant gloves, uvex profabutyl B-05R
手套
These unsupported gloves made from 100% butyl rubber provide a good resistance to polar compounds such as esters, ketones, aldehydes, amines and saturated saline solutions, plus acids and alkalis.
  • Seamlessly coated with bromobutyl (approx. 0,50 mm)
  • Good grip in damp and wet areas
  • Highly flexible, even at low temperatures
  • Impermeable to water vapour, gases and toxic substances
Please note: Butyl is not resistant to oil, grease, aliphatic and aromatic hydrocarbons or chlorinated hydrocarbons.
Certifications: EN 388:2016 (2010X), EN ISO 374-1:2016More Product Information