Crucibles, porcelain
供应商: USBECK LABORGERATE
5665 5666 5667 5668
USBE5665EA 0 CNY
USBE5665 USBE5666 USBE5667 USBE5668
Crucibles, porcelain
坩埚
These crucibles are made from glazed porcelain.
  • Temperature resistant up to 1000 °C
Certifications: DIN 12904