Chemist's Spoon
供应商: USBECK LABORGERATE
3406
USBE3406EA 0 CNY
USBE3406
Chemist's Spoon
勺子
Single-ended chemical spoon made from stainless steel.
  • Autoclavable
  • Easy to clean
  • 16 mm width
  • 12 g weight
Excellent chemical resistance. High temperature resistance.