Bossheads, globular
供应商: USBECK LABORGERATE
2360 2362 2363
USBE2360EA 0 CNY
USBE2360 USBE2362 USBE2363
Bossheads, globular
夹具 锤头
Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
  • 35 mm Ø
  • For rods up to 13 mm Ø
  • Nickel plated hinged screws
Weight: 100 g