Vacuum pumps, deep vacuum, oil
供应商: THERMO SAVANT
VLP285-230
THSAVLP285-230EA 0 CNY
THSAVLP285-230
Vacuum pumps, deep vacuum, oil
真空转子蒸发器泵
Deep vacuum oil pump is designed to work with SpeedVac applications.
  • Three-position gas ballast control
  • Standard oil drain kit simplifies oil change
  • Two separate oil inlet ports
  • Recommended for applications requiring high vacuum
订购须知 Includes oil mist eliminator (OME190), 2 filters, and line cord.