Drying ovens, Heratherm® advanced protocol security series
供应商: Thermo Fisher Scientific
51028154 51028155 51028156 51028545 51028145 51028146 51028147 51028546 51028547 51028548 51028549 51028544
THBR51028154EA 0 CNY
THBR51028154 THBR51028155 THBR51028156 THBR51028545 THBR51028145 THBR51028146 THBR51028147 THBR51028546 THBR51028547 THBR51028548 THBR51028549 THBR51028544
Drying ovens, Heratherm® advanced protocol security series
干燥箱 风扇辅助和自然对流炉
Heating and drying ovens with coated or stainless steel exterior. The timer allows the oven to be turned on or off after a preset time, with a choice between weekly, hourly or real time settings. Units with natural convection are ideal for gentle drying and heating, to protect delicate samples. Units with forced convection have adjustable fan speed and are suitable for faster drying and heating, and superior temperature uniformity.
  • Standard over-temperature alarm and additional under-temperature alarm
  • Large, easy to read vacuum fluorescent display and
More Product Information