Small compressed head swab, CleanFoam®
供应商: ITW-CONTAMINATION TEXWIPE
TX751B
TEXWTX751BEA 0 CNY
TEXWTX751B
Small compressed head swab, CleanFoam®
拭子 清洗用拭子
Excellent swab for cleaning very small areas, especially thin, grooved areas of parts and assemblies.
  • Very low particle generation
  • Small compressed CleanFoam head with pointed tip on a short rigid handle
  • Polyurethane foam head has been solvent washed to greatly reduce non volatile residue
Suitable for precision application of solvents and cleaning solutions.
Packaging: Cleanroom processed bag-within-a-bag.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904