New fuchsine for biochemical research

供应商: TCI
Warning

Synonyms: Basic Violet 2, New Fuchsin, 4-[(4-Amino-m-tolyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

N0873-5G
TCIAN0873-5GEA 0 CNY
TCIAN0873-5G
New fuchsine for biochemical research
New fuchsine
分子式: C₂₂H₂₄ClN₃
分子量: 365.91 g/mol
存储条件: Ambient
MDL编号: MFCD00012567
CAS号: 3248-91-7
EINECS: 221-831-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c596135365e2ac71763e0ecfdb89cf89e87c044c&vid=6fcfc086262bc7d3edc9972607a78219df7eaaa3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Green to Dark green powder to crystal
Lambda max. 550.0 to 558.0 nm (EtOH)
Absorbance (E1%1cm) min. 1,500 (EtOH, 550.0 to 558.0 nm)
Alkaline Phosphatase detection test to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR