2-Isobutyl-4-methylthiazole ≥96.0% (by GC)

供应商: TCI

I0442-5ML
TCIAI0442-5MLEA 0 CNY
TCIAI0442-5ML
2-Isobutyl-4-methylthiazole ≥96.0% (by GC)
2-Isobutyl-4-methylthiazole
分子式: C₈H₁₃NS
分子量: 155.2605 g/mol
沸点: 189°C
MDL编号: MFCD00053138
CAS号: 61323-24-8
EINECS: 262-709-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=059fdbb47a9adc3882e63ad156a5e41a2ab8d572&vid=059fdbb47a9adc3882e63ad156a5e41a2ab8d572&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 96.0 %

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR