Methyl-3,5-dibromo-4-hydroxybenzoate ≥99.0% (by GC)

供应商: TCI
Warning

D2501-25G
TCIAD2501-25GEA 0 CNY
TCIAD2501-25G
Methyl-3,5-dibromo-4-hydroxybenzoate ≥99.0% (by GC)
Methyl-3,5-dibromo-4-hydroxybenzoate
分子式: C₈H₆Br₂O₃
分子量: 309.94 g/mol
MDL编号: MFCD00016416
CAS号: 41727-47-3
EINECS: 255-521-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea66422de32d24f499033a65c065a4b3b9b648f5&vid=ea66422de32d24f499033a65c065a4b3b9b648f5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 122.0 to 125.0 deg-C
Solubility in Methanol within very faint turbidity
Water 5.0 to 6.0 %

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR