1,5-二苯基卡巴肼 98,0 HPLC_ASSAY_METHOD

供应商: TCI
Warning

Synonyms: 1,5-二苯羰酰肼, 对称二苯氨基脲

D1513-25G
TCIAD1513-25GEA 0 CNY
TCIAD1513-25G
1,5-二苯基卡巴肼 98,0 HPLC_ASSAY_METHOD
1,5-二苯基卡巴肼
分子式: C₁₃H₁₄N₄O
分子量: 242.28 g/mol
沸点: 454
存储条件: Freezer
MDL编号: MFCD00003013
CAS号: 140-22-7
EINECS: 205-403-7
Merck Index: 14,03321
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=603f15fc1192269743f3751d214b26da9a518cb6&vid=603f15fc1192269743f3751d214b26da9a518cb6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Solubility in Acetone almost transparency
Suitability for chrome-analysis Abs min.0.42 (near 540nm)in the presence of Dichromate (1 ppm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR